Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291824.1 | NP_001278753.1 | 7 | 987 | ||
NM_001291825.1 | NP_001278754.1 | 6 | 1000 | |||
NM_001291826.1 | NP_001278755.1 | 6 | 895 | |||
NM_001291828.1 | NP_001278757.1 | 4 | 597 | |||
NM_004810.3 | NP_004801.1 | 7 | 973 | |||
XM_006724376.3 | XP_006724439.1 | 6 | 890 | |||
GenBank mRNA | AF042380.1 | - | 7 | 902 | ||
AF090456.1 | - | 7 | 864 | |||
AF102694.1 | - | 7 | 902 | |||
AF129476.1 | - | 7 | 892 | |||
AF236119.1 | - | 7 | 933 | |||
AF236120.1 | - | 6 | 710 | |||
AJ011736.1 | - | 7 | 918 | |||
AK303280.1 | - | 6 | 880 | |||
AK303332.1 | - | 5 | 787 | |||
AK303470.1 | - | 4 | 588 | |||
AK303512.1 | - | 4 | 642 | |||
AK303657.1 | - | 6 | 852 | |||
AK312830.1 | - | 7 | 972 | |||
AY069959.1 | - | 7 | 973 | |||
BC025692.1 | - | 7 | 907 | 5208327 | ||
BC026002.1 | - | 6 | 969 | 4672797 | ||
CR456498.1 | - | 7 | 771 | |||
CR536524.1 | - | 6 | 710 | |||
CR541849.1 | - | 6 | 710 |