Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143995.2 | NP_001137467.1 | 2, 3 | 389 | ||
NM_004811.2 | NP_004802.1 | 2, 3 | 326 | |||
XM_006718750.1 | XP_006718813.1 | 2, 3 | 422 | |||
XM_011545393.2 | XP_011543695.1 | 2, 3 | 369 | |||
XM_011545394.2 | XP_011543696.1 | 2, 3 | 292 | |||
XM_017018579.1 | XP_016874068.1 | 2, 3 | 369 | |||
GenBank mRNA | AF062075.1 | - | 2, 3 | 274 | ||
AK223165.1 | - | 2, 3 | 265 | |||
AK300955.1 | - | 2, 3 | 316 | |||
AK301093.1 | - | 2, 3 | 265 | |||
AK313306.1 | - | 2, 3 | 259 | |||
BC019035.2 | - | 2, 3 | 248 | 4575069 | ||
BP261314.1 | - | 2, 3 | 326 | |||
CR536531.1 | - | 1, 2 | 181 | |||
DA363664.1 | - | 2, 3 | 423 |