Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS11D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004262.2 | NP_004253.1 | 7 | 707 | ||
XM_005265710.4 | XP_005265767.1 | 6 | 1650 | |||
XM_017008851.1 | XP_016864340.1 | 3 | 170 | |||
GenBank mRNA | AB002134.1 | - | 7 | 703 | ||
AK094054.1 | - | 1 | 374 | |||
AK290289.1 | - | 7 | 708 | |||
AK296853.1 | - | 6 | 570 | |||
BC125195.1 | - | 7 | 692 | 40123006 | ||
BC125196.1 | - | 7 | 692 | 40123011 |