Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271811.1 | NP_001258740.1 | 1, 2 | 95 | ||
NM_004264.4 | NP_004255.2 | 1, 2 | 95 | |||
GenBank mRNA | AK311838.1 | - | 1, 2 | 39 | ||
BC008380.1 | - | 1, 2 | 67 | 4041267 | ||
CR456999.1 | - | 1, 2 | 37 | |||
DB024509.1 | - | 1, 2 | 95 | |||
HY075642.1 | - | 1, 2 | 69 | |||
KU178474.1 | - | 1, 2 | 37 | |||
KU178475.1 | - | 1, 2 | 37 | |||
U46837.1 | - | 1, 2 | 46 | |||
U52960.1 | - | 1, 2 | 42 |