Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TJP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170414.2 | NP_001163885.1 | 16 | 2414 | ||
NM_001170415.1 | NP_001163886.1 | 15 | 2414 | |||
NM_001170416.1 | NP_001163887.1 | 15 | 2344 | |||
NM_004817.3 | NP_004808.2 | 15 | 2569 | |||
NM_201629.3 | NP_963923.1 | 15 | 2569 | |||
XM_005252314.2 | XP_005252371.1 | 15 | 2622 | |||
XM_011519204.1 | XP_011517506.1 | 15 | 2312 | |||
XM_011519206.2 | XP_011517508.1 | 17 | 2914 | |||
XM_011519207.2 | XP_011517509.1 | 16 | 2410 | |||
XM_011519208.2 | XP_011517510.1 | 16 | 2370 | |||
XM_011519209.2 | XP_011517511.1 | 14 | 2199 | |||
XM_017015326.1 | XP_016870815.1 | 16 | 2467 | |||
XM_017015327.1 | XP_016870816.1 | 15 | 2569 | |||
XM_017015328.1 | XP_016870817.1 | 15 | 2622 | |||
GenBank mRNA | AF489824.1 | - | 15 | 2330 | ||
AK295016.1 | - | 16 | 2370 | |||
AK295034.1 | - | 15 | 2344 | |||
AK302483.1 | - | 15 | 2270 | |||
AK304447.1 | - | 17 | 2496 | |||
BC027592.1 | - | 15 | 2460 | 4839233 | ||
L27476.1 | - | 15 | 2330 |