Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 5 | 656 | ||
NM_001281502.1 | NP_001268431.1 | 5 | 725 | |||
NM_004265.3 | NP_004256.1 | 5 | 997 | |||
GenBank mRNA | AF084559.1 | - | 5 | 789 | ||
AF108658.1 | - | 5 | 582 | |||
AF126799.1 | - | 5 | 652 | |||
AK074925.1 | - | 3 | 204 | |||
AK074939.1 | - | 5 | 729 | |||
AK074991.1 | - | 5 | 465 | |||
AK290291.1 | - | 5 | 603 | |||
AK299762.1 | - | 5 | 725 | |||
AL050118.1 | - | 3 | 241 | |||
BC009011.1 | - | 5 | 731 | 4181037 |