Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004823.1 | NP_004814.1 | 2 | 713 | ||
XM_011527526.1 | XP_011525828.1 | 3 | 1275 | |||
GenBank mRNA | AF117708.1 | - | 2 | 713 | ||
AF134149.1 | - | 2 | 722 | |||
AF281302.1 | - | 2 | 678 | |||
AF281303.1 | - | 2 | 302 | |||
AK022344.1 | - | 2 | 442 | |||
AK222545.1 | - | 2 | 695 | |||
AK315653.1 | - | 2 | 687 | |||
BC004367.1 | - | 2 | 643 | 3639657 | ||
EU978937.1 | - | 2 | 661 |