Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021999.4 | NP_068839.1 | 2 | 397 | ||
GenBank mRNA | AF086918.1 | - | 2 | 289 | ||
AF092128.1 | - | 2 | 386 | |||
AF136973.1 | - | 2 | 386 | |||
AF152462.1 | - | 2 | 344 | |||
AF246221.1 | - | 2 | 344 | |||
AK000503.1 | - | 2 | 329 | |||
AK027411.1 | - | 2 | 329 | |||
AK130048.1 | - | 2 | 333 | |||
BC000554.2 | - | 2 | 323 | 3163436 | ||
BC016148.1 | - | 2 | 344 | 3912066 | ||
BT006863.1 | - | 2 | 174 | |||
DA087198.1 | - | 2 | 397 |