Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004833.1 | NP_004824.1 | 3 | 547 | ||
XM_005245616.4 | XP_005245673.1 | 2 | 318 | |||
XM_017002848.1 | XP_016858337.1 | 4 | 1774 | |||
GenBank mRNA | AF024714.1 | - | 3 | 547 | ||
AK292042.1 | - | 3 | 456 | |||
BC010940.1 | - | 3 | 549 | 4080360 |