Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
siRNA ID
HSS114052
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
21
2456
NM_001242560.1
NP_001229489.1
22
2817
NM_004834.4
NP_004825.3
21
2583
NM_145686.3
NP_663719.2
21
2739
NM_145687.3
NP_663720.1
22
2375
XM_005264048.2
XP_005264105.1
23
2768
XM_005264050.3
XP_005264107.1
22
2606
XM_005264052.3
XP_005264109.1
22
2603
XM_005264057.3
XP_005264114.1
22
2537
XM_005264058.3
XP_005264115.1
21
2528
XM_005264059.3
XP_005264116.1
22
2444
XM_005264060.3
XP_005264117.1
21
2432
XM_005264062.3
XP_005264119.1
21
2375
XM_005264063.3
XP_005264120.1
21
2372
XM_005264064.3
XP_005264121.1
20
2366
XM_005264065.3
XP_005264122.1
21
2282
XM_005264066.3
XP_005264123.1
21
2279
XM_005264068.3
XP_005264125.1
20
2273
XM_005264069.2
XP_005264126.1
21
2435
XM_005264071.2
XP_005264128.1
20
2363
XM_006712865.2
XP_006712928.1
22
2513
XM_006712868.2
XP_006712931.1
21
2282
XM_006712869.2
XP_006712932.1
21
2375
XM_017005349.1
XP_016860838.1
23
2672
XM_017005350.1
XP_016860839.1
22
2606
XM_017005351.1
XP_016860840.1
21
2525
XM_017005352.1
XP_016860841.1
22
2537
XM_017005353.1
XP_016860842.1
22
2534
XM_017005354.1
XP_016860843.1
22
2513
XM_017005355.1
XP_016860844.1
21
2504
XM_017005356.1
XP_016860845.1
21
2435
XM_017005357.1
XP_016860846.1
22
2444
XM_017005358.1
XP_016860847.1
22
2441
XM_017005359.1
XP_016860848.1
22
2606
XM_017005360.1
XP_016860849.1
22
2603
XM_017005361.1
XP_016860850.1
22
2606
XM_017005362.1
XP_016860851.1
22
2603
XM_017005363.1
XP_016860852.1
21
2375
XM_017005364.1
XP_016860853.1
20
2363
XM_017005365.1
XP_016860854.1
22
2537
XM_017005366.1
XP_016860855.1
22
2513
XM_017005367.1
XP_016860856.1
22
2510
XM_017005368.1
XP_016860857.1
22
2441
XM_017005369.1
XP_016860858.1
21
2372
XM_017005370.1
XP_016860859.1
21
2372
XM_017005371.1
XP_016860860.1
20
2366
XM_017005372.1
XP_016860861.1
21
2282
XM_017005373.1
XP_016860862.1
20
2273
XM_017005374.1
XP_016860863.1
21
2282
XM_017005375.1
XP_016860864.1
20
2273
GenBank mRNA
AB013385.1
-
5
363
AB014587.1
-
20
2259
AF096300.1
-
21
2284
AK001272.1
-
5
518
AK025610.1
-
9
1132
AK295610.1
-
20
2409
AK296396.1
-
22
2513
AL137755.1
-
14
1912
AY212247.1
-
21
2451
AY834276.1
-
17
1860
EU794634.1
-
22
2425
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0