Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277077.1 | NP_001264006.1 | 3 | 1337 | ||
NM_001277078.1 | NP_001264007.1 | 3 | 1337 | |||
NM_004272.4 | NP_004263.1 | 3 | 1337 | |||
XM_017010059.1 | XP_016865548.1 | 3 | 2069 | |||
GenBank mRNA | AF093262.1 | - | 2 | 275 | ||
AK223471.1 | - | 3 | 599 | |||
AK312481.1 | - | 3 | 602 | |||
AY189939.1 | - | 3 | 295 | |||
AY189941.1 | - | 3 | 295 | |||
BC015502.1 | - | 3 | 552 | 3904242 | ||
BC047438.1 | - | 3 | 1337 | 5294849 | ||
BT009846.1 | - | 2 | 275 | |||
Y17829.1 | - | 3 | 350 |