Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321643.1 | NP_001308572.1 | 25 | 3066 | ||
NM_004850.4 | NP_004841.2 | 25 | 3522 | |||
XM_005246190.3 | XP_005246247.1 | 25 | 3556 | |||
XM_011510417.2 | XP_011508719.1 | 25 | 3021 | |||
XM_017005378.1 | XP_016860867.1 | 25 | 3553 | |||
XM_017005379.1 | XP_016860868.1 | 25 | 2951 | |||
GenBank mRNA | AB014519.1 | - | 25 | 3527 | ||
AK299296.1 | - | 11 | 1529 | |||
AK310358.1 | - | 21 | 2817 | |||
BC111801.1 | - | 12 | 1629 | 40069866 | ||
D87931.1 | - | 25 | 3073 |