Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204051.1 | NP_001190980.1 | 2 | 531 | ||
NM_001204052.1 | NP_001190981.1 | 2 | 531 | |||
NM_004277.4 | NP_004268.3 | 2 | 531 | |||
XM_005249483.2 | XP_005249540.1 | 2 | 501 | |||
XM_017011512.1 | XP_016867001.1 | 1 | 384 | |||
GenBank mRNA | AF110532.1 | - | 2 | 253 | ||
AK291427.1 | - | 2 | 461 | |||
AK295233.1 | - | 2 | 514 | |||
AK302906.1 | - | 2 | 471 | |||
AK315940.1 | - | 2 | 442 | |||
AY358711.1 | - | 2 | 279 | |||
BC033091.1 | - | 2 | 504 | 5455903 | ||
BP310559.1 | - | 3 | 418 | |||
DA144842.1 | - | 2 | 505 | |||
DC314392.1 | - | 2 | 531 | |||
DR002070.1 | - | 2 | 453 |