Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204051.1 | NP_001190980.1 | 3 | 594 | ||
NM_001204052.1 | NP_001190981.1 | 3 | 594 | |||
NM_004277.4 | NP_004268.3 | 3 | 594 | |||
XM_005249483.2 | XP_005249540.1 | 3 | 564 | |||
XM_017011512.1 | XP_016867001.1 | 2 | 443 | |||
GenBank mRNA | AF110532.1 | - | 3 | 316 | ||
AK090871.1 | - | 1 | 923 | |||
AK291427.1 | - | 3 | 524 | |||
AK295233.1 | - | 3 | 577 | |||
AK302906.1 | - | 4 | 634 | |||
AK315940.1 | - | 3 | 505 | |||
AY358711.1 | - | 3 | 342 | |||
BC033091.1 | - | 3 | 567 | 5455903 | ||
BP310559.1 | - | 4 | 481 | |||
DR002070.1 | - | 3 | 516 |