Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004278.3 | NP_004269.1 | 5 | 543 | ||
XM_017025349.1 | XP_016880838.1 | 5 | 529 | |||
XM_017025350.1 | XP_016880839.1 | 5 | 529 | |||
XM_017025351.1 | XP_016880840.1 | 5 | 529 | |||
XM_017025352.1 | XP_016880841.1 | 5 | 529 | |||
XM_017025353.1 | XP_016880842.1 | 5 | 529 | |||
XM_017025356.1 | XP_016880845.1 | 5 | 529 | |||
GenBank mRNA | AB017165.1 | - | 5 | 516 | ||
AK292932.1 | - | 5 | 528 | |||
AK296120.1 | - | 4 | 418 | |||
AK302523.1 | - | 5 | 528 | |||
BC068197.1 | - | 5 | 522 | 6711166 |