Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK303874.1 | - | 10 | 1056 | ||
AK308322.1 | - | 13 | 1447 | |||
AK308361.1 | - | 8 | 871 | |||
BC017705.2 | - | 13 | 1430 | |||
BC051826.1 | - | 14 | 1604 | 6500787 | ||
X67155.2 | - | 13 | 1502 |