Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004857.3 | NP_004848.3 | 2 | 694 | ||
GenBank mRNA | AK315336.1 | - | 2 | 695 | ||
BC057229.1 | - | 2 | 608 | |||
BC131516.1 | - | 1 | 926 | 40084320 | ||
M90359.1 | - | 1 | 1613 |