Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321120.1 | NP_001308049.1 | 9 | 1416 | ||
NM_018488.3 | NP_060958.2 | 8 | 1253 | |||
XM_011525490.2 | XP_011523792.1 | 9 | 1454 | |||
XM_011525491.2 | XP_011523793.1 | 9 | 1451 | |||
GenBank mRNA | AF188703.1 | - | 8 | 1295 | ||
BC136403.1 | - | 9 | 1277 | 9020390 | ||
BC142620.1 | - | 9 | 1416 | 40147430 | ||
BC144062.1 | - | 9 | 1280 | 9052582 |