Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005332.1 | NP_001005332.1 | 12 | 2177 | ||
NM_001005333.1 | NP_001005333.1 | 13 | 2440 | |||
NM_006986.3 | NP_008917.3 | 12 | 2272 | |||
XM_011530835.2 | XP_011529137.1 | 13 | 2295 | |||
XM_017029978.1 | XP_016885467.1 | 12 | 2187 | |||
XM_017029979.1 | XP_016885468.1 | 13 | 2873 | |||
XM_017029980.1 | XP_016885469.1 | 13 | 2691 | |||
GenBank mRNA | AF124440.1 | - | 10 | 1479 | ||
AF132205.1 | - | 9 | 899 | |||
AF217963.1 | - | 12 | 2222 | |||
AF258554.1 | - | 12 | 2246 | |||
AF300328.1 | - | 12 | 2223 | |||
AK074092.1 | - | 6 | 4739 | |||
AK314053.1 | - | 10 | 2131 | |||
AL133628.1 | - | 9 | 1177 | |||
BC014070.2 | - | 13 | 2403 | 4026470 | ||
BC032473.2 | - | 11 | 2119 | 5216354 |