Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004863.3 | NP_004854.1 | 6 | 1006 | ||
XM_011537384.2 | XP_011535686.1 | 6 | 902 | |||
GenBank mRNA | AB011098.1 | - | 6 | 1006 | ||
BC005123.2 | - | 6 | 907 | 3688136 | ||
BQ338973.1 | - | 3 | 338 | |||
U15555.1 | - | 5 | 642 | |||
Y08686.1 | - | 6 | 866 |