Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004864.2 | NP_004855.2 | 1 | 52 | ||
GenBank mRNA | AK291530.1 | - | 1 | 52 | ||
BC000529.2 | - | 1 | 31 | 2961614 | ||
BC008962.2 | - | 1 | 37 | 3009858 | ||
U88323.1 | - | 1 | 45 |