Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004864.2 | NP_004855.2 | 2 | 538 | ||
GenBank mRNA | AB000584.1 | - | 2 | 538 | ||
AF003934.1 | - | 2 | 540 | |||
AF019770.1 | - | 2 | 523 | |||
AK225431.1 | - | 2 | 538 | |||
AK291530.1 | - | 2 | 538 | |||
BC000529.2 | - | 2 | 517 | 2961614 | ||
BC008962.2 | - | 2 | 523 | 3009858 | ||
BT019465.1 | - | 2 | 506 | |||
U51731.1 | - | 2 | 518 | |||
U88323.1 | - | 2 | 531 |