Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004866.5 | NP_004857.4 | 2 | 269 | ||
NR_110885.1 | 2 | 269 | ||||
XM_011543727.2 | XP_011542029.1 | 2 | 269 | |||
GenBank mRNA | AF005037.1 | - | 2 | 143 | ||
AF038966.1 | - | 2 | 190 | |||
AK225172.1 | - | 2 | 158 | |||
AK290225.1 | - | 2 | 119 | |||
AK298146.1 | - | 2 | 139 | |||
BC009787.1 | - | 2 | 161 | |||
BC015065.1 | - | 2 | 162 | 4105068 | ||
BC034048.1 | - | 2 | 123 | 4814951 | ||
CB960371.1 | - | 269 | ||||
CR542286.1 | - | 2 | 100 | |||
DB213762.1 | - | 2 | 269 |