Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015048.2 | NP_001015048.1 | 2 | 1251 | ||
NM_001015049.2 | NP_001015049.1 | 2 | 1458 | |||
NM_004873.3 | NP_004864.1 | 2 | 1271 | |||
GenBank mRNA | AB020680.1 | - | 2 | 1081 | ||
AB105186.1 | - | 2 | 1247 | |||
AF095195.2 | - | 2 | 1247 | |||
AK001378.1 | - | 1 | 309 | |||
AK023145.1 | - | 1 | 778 | |||
AK225730.1 | - | 2 | 1246 | |||
AK225741.1 | - | 2 | 1219 | |||
AK291312.1 | - | 2 | 1190 | |||
BC044216.1 | - | 2 | 1417 | 5745133 | ||
BC050551.1 | - | 2 | 1252 | 5744362 |