Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015048.2 | NP_001015048.1 | 2 | 601 | ||
NM_001015049.2 | NP_001015049.1 | 2 | 808 | |||
NM_004873.3 | NP_004864.1 | 2 | 621 | |||
GenBank mRNA | AB020680.1 | - | 2 | 431 | ||
AB105186.1 | - | 2 | 597 | |||
AF095195.2 | - | 2 | 597 | |||
AK023145.1 | - | 1 | 128 | |||
AK225730.1 | - | 2 | 596 | |||
AK225741.1 | - | 2 | 569 | |||
AK291312.1 | - | 2 | 540 | |||
BC044216.1 | - | 2 | 767 | 5745133 | ||
BC050551.1 | - | 2 | 602 | 5744362 |