Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 6 | 575 | ||
NM_004879.4 | NP_004870.3 | 6 | 617 | |||
NR_110769.1 | 5 | 471 | ||||
NR_110770.1 | 7 | 742 | ||||
XM_011543069.1 | XP_011541371.1 | 7 | 633 | |||
XM_011543070.1 | XP_011541372.1 | 6 | 788 | |||
XM_017018582.1 | XP_016874071.1 | 7 | 633 | |||
GenBank mRNA | AB209820.1 | - | 6 | 574 | ||
AK223029.1 | - | 6 | 546 | |||
AK291951.1 | - | 6 | 526 | |||
AK296620.1 | - | 6 | 504 | |||
AK301556.1 | - | 5 | 400 | |||
AK315841.1 | - | 6 | 533 | |||
AK316539.1 | - | 7 | 652 | |||
BC002390.2 | - | 6 | 483 | 2819783 | ||
BG566671.1 | - | 6 | 593 |