Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004286.4 | NP_004277.2 | 8 | 1405 | ||
XM_011530537.2 | XP_011528839.1 | 8 | 2667 | |||
XM_017029099.1 | XP_016884588.1 | 8 | 2659 | |||
XM_017029100.1 | XP_016884589.1 | 8 | 2629 | |||
XM_017029101.1 | XP_016884590.1 | 9 | 1246 | |||
XM_017029102.1 | XP_016884591.1 | 7 | 1062 | |||
XM_017029103.1 | XP_016884592.1 | 7 | 1160 | |||
GenBank mRNA | AK315704.1 | - | 7 | 1028 | ||
BC014075.1 | - | 8 | 1243 | |||
CR456501.1 | - | 8 | 1228 | |||
U87964.1 | - | 8 | 1149 |