Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004286.4 | NP_004277.2 | 11 | 1906 | ||
XM_011530537.2 | XP_011528839.1 | 11 | 3168 | |||
XM_017029099.1 | XP_016884588.1 | 11 | 3160 | |||
XM_017029100.1 | XP_016884589.1 | 11 | 3130 | |||
XM_017029101.1 | XP_016884590.1 | 12 | 1747 | |||
XM_017029102.1 | XP_016884591.1 | 10 | 1563 | |||
XM_017029103.1 | XP_016884592.1 | 10 | 1661 | |||
GenBank mRNA | AK096647.1 | - | 1 | 1242 | ||
AK315704.1 | - | 10 | 1529 | |||
BC014075.1 | - | 11 | 1744 | |||
CR456501.1 | - | 11 | 1729 | |||
U87964.1 | - | 11 | 1650 |