Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253854.1 | NP_001240783.1 | 9, 10 | 1380 | ||
NM_001253855.1 | NP_001240784.1 | 5, 6 | 1111 | |||
NM_012443.3 | NP_036575.1 | 6, 7 | 1025 | |||
NM_172242.2 | NP_758442.1 | 6, 7 | 1025 | |||
XM_005252645.1 | XP_005252702.1 | 6, 7 | 1054 | |||
GenBank mRNA | AF079363.1 | - | 6, 7 | 982 | ||
AK289903.1 | - | 6, 7 | 974 | |||
AK302194.1 | - | 9, 10 | 1355 | |||
AK310247.1 | - | 5, 6 | 1064 | |||
AL080136.1 | - | 6, 7 | 973 | |||
BC030585.2 | - | 6, 7 | 973 | 4822996 | ||
CR533552.1 | - | 6, 7 | 858 |