Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128930.2 | NP_001122402.1 | 12 | 1738 | ||
NM_004901.4 | NP_004892.1 | 12 | 1762 | |||
GenBank mRNA | AB002390.1 | - | 10 | 1292 | ||
AF016032.1 | - | 12 | 1667 | |||
AJ131358.1 | - | 12 | 1659 | |||
AK300464.1 | - | 12 | 1670 | |||
BC034477.2 | - | 12 | 1702 | 4824559 | ||
DB091432.1 | - | 4 | 482 |