Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128930.2 | NP_001122402.1 | 13 | 1927 | ||
NM_004901.4 | NP_004892.1 | 13 | 1951 | |||
GenBank mRNA | AB002390.1 | - | 11 | 1481 | ||
AF016032.1 | - | 13 | 1856 | |||
AJ131358.1 | - | 13 | 1848 | |||
CN313713.1 | - | 2 | 167 |