Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003690.1 | NP_001003690.1 | 3 | 574 | ||
NM_014628.2 | NP_055443.1 | 2 | 306 | |||
GenBank mRNA | AK297172.1 | - | 2 | 327 | ||
AL537692.3 | - | 3 | 576 | |||
BC002904.2 | - | 2 | 276 | 3945348 | ||
BE297042.1 | - | 3 | 502 | |||
BI255998.1 | - | 2 | 293 | |||
BU845738.1 | - | 3 | 611 | |||
CB991410.1 | - | 2 | 333 | |||
CR456893.1 | - | 2 | 270 | |||
D14811.1 | - | 2 | 273 |