Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004911.4 | NP_004902.1 | 7 | 1305 | ||
XM_006716185.2 | XP_006716248.1 | 7 | 1305 | |||
GenBank mRNA | AK290971.1 | - | 7 | 1090 | ||
AK313299.1 | - | 7 | 1277 | |||
BC000425.2 | - | 7 | 1067 | 2819726 | ||
BC001928.1 | - | 7 | 1067 | 2819726 | ||
BC006344.2 | - | 7 | 1255 | 4098367 | ||
BC011754.2 | - | 7 | 1256 | 3532913 |