Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004914.3 | NP_004905.2 | 5 | 520 | ||
XM_005261859.4 | XP_005261916.1 | 6 | 604 | |||
XM_006724381.3 | XP_006724444.1 | 6 | 607 | |||
XM_006724382.3 | XP_006724445.1 | 5 | 532 | |||
XM_011530544.2 | XP_011528846.1 | 6 | 612 | |||
XM_011530545.2 | XP_011528847.1 | 6 | 608 | |||
XM_011530546.2 | XP_011528848.1 | 6 | 614 | |||
XM_011530547.2 | XP_011528849.1 | 4 | 454 | |||
XM_011530550.2 | XP_011528852.1 | 6 | 618 | |||
XM_017029104.1 | XP_016884593.1 | 5 | 528 | |||
XM_017029105.1 | XP_016884594.1 | 4 | 450 | |||
XM_017029106.1 | XP_016884595.1 | 4 | 332 | |||
XM_017029107.1 | XP_016884596.1 | 5 | 368 | |||
GenBank mRNA | AB023061.1 | - | 5 | 520 | ||
AF133588.1 | - | 5 | 490 | |||
AY336745.1 | - | 4 | 414 | |||
BC104989.1 | - | 5 | 509 | 8143992 | ||
BC104991.1 | - | 5 | 509 | 8143994 | ||
CR456553.1 | - | 5 | 498 |