Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004292.2 | NP_004283.2 | 10 | 2384 | ||
XM_011545399.2 | XP_011543701.1 | 10 | 2733 | |||
XM_011545400.2 | XP_011543702.1 | 10 | 2103 | |||
XM_011545401.2 | XP_011543703.1 | 10 | 2258 | |||
XM_017018587.1 | XP_016874076.1 | 10 | 2106 | |||
GenBank mRNA | AB209445.1 | - | 7 | 2287 | ||
AK295365.1 | - | 9 | 1975 | |||
AK301430.1 | - | 7 | 1673 | |||
BC014417.1 | - | 10 | 2274 | 4549333 | ||
BG163919.1 | - | 1 | 165 | |||
KU178481.1 | - | 10 | 2257 | |||
KU178482.1 | - | 10 | 2071 | |||
KU178483.1 | - | 4 | 524 | |||
L36463.1 | - | 10 | 2384 |