Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004292.2 | NP_004283.2 | 2, 3 | 403 | ||
XM_011545399.2 | XP_011543701.1 | 2, 3 | 752 | |||
XM_011545400.2 | XP_011543702.1 | 2, 3 | 308 | |||
XM_011545401.2 | XP_011543703.1 | 2, 3 | 277 | |||
XM_017018587.1 | XP_016874076.1 | 2, 3 | 308 | |||
GenBank mRNA | AB209445.1 | - | 1, 2 | 40 | ||
BC014417.1 | - | 2, 3 | 293 | 4549333 | ||
KU178481.1 | - | 2, 3 | 276 | |||
KU178482.1 | - | 2, 3 | 276 | |||
L36463.1 | - | 2, 3 | 403 |