Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOR1 SIRNA Assays ›
Gene Symbol:
NCOR1
Gene Name:
nuclear receptor corepressor 1
Gene Aliases:
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location:
Chr.17: 16030094 - 16216746 on Build GRCh38
UniGene ID:
Hs.462323
Species:
Human
siRNA ID
HSS114353
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NCOR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190438.1
NP_001177367.1
18
2566
NM_001190440.1
NP_001177369.1
20
2893
NM_006311.3
NP_006302.2
21
3044
XM_005256866.4
XP_005256923.1
22
3105
XM_005256868.4
XP_005256925.1
22
3102
XM_005256871.4
XP_005256928.1
21
3078
XM_005256872.4
XP_005256929.1
21
3075
XM_005256873.4
XP_005256930.1
22
3057
XM_005256874.4
XP_005256931.1
22
3105
XM_005256875.3
XP_005256932.1
22
3105
XM_006721601.3
XP_006721664.1
23
3237
XM_006721602.3
XP_006721665.1
23
3232
XM_006721603.3
XP_006721666.1
22
3105
XM_006721604.3
XP_006721667.1
21
3075
XM_011524083.2
XP_011522385.1
22
4016
XM_011524084.2
XP_011522386.1
22
3105
XM_011524085.2
XP_011522387.1
22
3105
XM_011524086.2
XP_011522388.1
21
3075
XM_017025396.1
XP_016880885.1
23
3982
XM_017025397.1
XP_016880886.1
23
3747
XM_017025398.1
XP_016880887.1
22
3189
XM_017025399.1
XP_016880888.1
22
3208
XM_017025400.1
XP_016880889.1
23
3979
XM_017025401.1
XP_016880890.1
23
3744
XM_017025402.1
XP_016880891.1
22
3028
XM_017025403.1
XP_016880892.1
22
3205
XM_017025404.1
XP_016880893.1
22
3696
XM_017025405.1
XP_016880894.1
21
3591
XM_017025406.1
XP_016880895.1
21
3989
XM_017025407.1
XP_016880896.1
21
2925
XM_017025408.1
XP_016880897.1
21
2997
XM_017025409.1
XP_016880898.1
22
3202
XM_017025410.1
XP_016880899.1
22
3698
XM_017025411.1
XP_016880900.1
21
3441
XM_017025412.1
XP_016880901.1
21
3218
XM_017025413.1
XP_016880902.1
21
3001
XM_017025414.1
XP_016880903.1
20
2908
XM_017025415.1
XP_016880904.1
22
3105
XM_017025416.1
XP_016880905.1
22
3713
XM_017025417.1
XP_016880906.1
22
3057
XM_017025418.1
XP_016880907.1
22
3105
XM_017025419.1
XP_016880908.1
22
3105
XM_017025420.1
XP_016880909.1
22
3057
GenBank mRNA
AB028970.2
-
20
2867
AF044209.1
-
21
3015
AF087856.1
-
18
2580
AF303586.1
-
18
2580
BC167431.1
-
20
2868
8860107
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
skeletal muscle tissue development
lactation
circadian rhythm
positive regulation of cell proliferation
covalent chromatin modification
cerebellum development
cellular lipid metabolic process
negative regulation of JNK cascade
spindle assembly
regulation of glycolytic process by negative regulation of transcription from RNA polymerase II promoter
regulation of lipid transport by negative regulation of transcription from RNA polymerase II promoter
negative regulation of production of miRNAs involved in gene silencing by miRNA
regulation of fatty acid transport
RNA polymerase II regulatory region DNA binding
RNA polymerase II activating transcription factor binding
chromatin binding
transcription corepressor activity
protein binding
ligand-dependent nuclear receptor binding
protein domain specific binding
estrogen receptor binding
nuclear hormone receptor binding
histone deacetylase binding
peroxisome proliferator activated receptor binding
transcription regulatory region DNA binding
retinoid X receptor binding
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline