Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077261.3 | NP_001070729.2 | 42, 43 | 6900 | ||
NM_001206654.1 | NP_001193583.1 | 42, 43 | 6900 | |||
NM_006312.5 | NP_006303.4 | 43, 44 | 6930 | |||
GenBank mRNA | AF113003.1 | - | 42, 43 | 6619 | ||
AF125672.1 | - | 41, 42 | 6744 | |||
AK127788.1 | - | 2, 3 | 1775 | |||
AY965853.1 | - | 41, 42 | 6744 | |||
BC004326.1 | - | 4, 5 | 885 | |||
S83390.1 | - | 9, 10 | 1967 |