Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004922.3 | NP_004913.2 | 13 | 1959 | ||
NM_198597.2 | NP_940999.1 | 12 | 1872 | |||
XM_011540380.2 | XP_011538682.1 | 13 | 1964 | |||
XM_011540381.2 | XP_011538683.1 | 14 | 2237 | |||
XM_017016967.1 | XP_016872456.1 | 14 | 1949 | |||
XM_017016968.1 | XP_016872457.1 | 13 | 2088 | |||
GenBank mRNA | AK298598.1 | - | 6 | 1059 | ||
AK299826.1 | - | 12 | 1778 | |||
AK309918.1 | - | 11 | 1709 | |||
AL832110.1 | - | 12 | 1854 | |||
BC018928.2 | - | 12 | 1860 | 4123623 | ||
D38555.1 | - | 12 | 1893 |