Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014630.2 | NP_055445.2 | 8 | 3331 | ||
XM_005254996.3 | XP_005255053.1 | 7 | 3232 | |||
XM_011522246.2 | XP_011520548.1 | 8 | 3401 | |||
XM_011522247.2 | XP_011520549.1 | 7 | 3404 | |||
XM_011522248.2 | XP_011520550.1 | 8 | 3362 | |||
XM_017022734.1 | XP_016878223.1 | 6 | 3112 | |||
GenBank mRNA | BC094688.1 | - | 8 | 3283 | 30397050 | |
BC112232.1 | - | 6 | 3065 | 8327700 | ||
BC112234.1 | - | 6 | 3066 | 8327702 | ||
BC143600.1 | - | 6 | 3056 | |||
BG757479.1 | - | 2 | 419 | |||
D86966.1 | - | 8 | 3565 |