Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162893.1 | NP_001156365.1 | 5 | 1090 | ||
NM_001162894.1 | NP_001156366.1 | 4 | 1013 | |||
NM_001162895.1 | NP_001156367.1 | 3 | 879 | |||
NM_001319230.1 | NP_001306159.1 | 3 | 939 | |||
NM_001319231.1 | NP_001306160.1 | 5 | 1028 | |||
NM_014656.2 | NP_055471.2 | 4 | 953 | |||
XM_017002909.1 | XP_016858398.1 | 4 | 1440 | |||
XM_017002910.1 | XP_016858399.1 | 4 | 1220 | |||
XM_017002911.1 | XP_016858400.1 | 4 | 1223 | |||
GenBank mRNA | AK292691.1 | - | 4 | 941 | ||
AL049799.1 | - | 4 | 933 | |||
AL049800.1 | - | 4 | 863 | |||
BC032361.1 | - | 4 | 871 | |||
BC111699.1 | - | 3 | 794 | |||
D25539.1 | - | 4 | 828 |