Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DEPDC5 SIRNA Assays ›
Gene Symbol:
DEPDC5
Gene Name:
DEP domain containing 5
Gene Aliases:
DEP.5, FFEVF
Chromosome Location:
Chr.22: 31753951 - 31907034 on Build GRCh38
UniGene ID:
Hs.435022
Species:
Human
siRNA ID
HSS114497
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DEPDC5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007188.2
NP_001007189.1
9
708
NM_001136029.2
NP_001129501.1
8
566
NM_001242896.1
NP_001229825.1
9
708
NM_001242897.1
NP_001229826.1
9
612
NM_014662.4
NP_055477.1
8
566
NR_110988.1
9
708
XM_005261862.1
XP_005261919.1
9
612
XM_011530557.2
XP_011528859.1
9
715
XM_011530558.2
XP_011528860.1
9
715
XM_011530559.2
XP_011528861.1
9
715
XM_011530560.2
XP_011528862.1
9
734
XM_011530561.2
XP_011528863.1
9
715
XM_011530562.2
XP_011528864.1
9
715
XM_011530563.2
XP_011528865.1
9
715
XM_011530565.2
XP_011528867.1
9
715
XM_011530568.2
XP_011528870.1
9
715
XM_017029109.1
XP_016884598.1
9
612
XM_017029110.1
XP_016884599.1
9
612
XM_017029111.1
XP_016884600.1
9
612
XM_017029112.1
XP_016884601.1
9
612
XM_017029113.1
XP_016884602.1
9
715
XM_017029114.1
XP_016884603.1
9
715
XM_017029115.1
XP_016884604.1
9
715
GenBank mRNA
AB014545.1
-
8
565
AJ698950.1
-
8
561
AJ698951.1
-
8
561
AJ704764.1
-
8
561
AK294987.1
-
9
584
AK300737.1
-
8
593
BC057797.1
-
9
648
BC136612.1
-
9
622
9020602
BC146766.1
-
8
565
9007209
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of TOR signaling
cellular response to amino acid starvation
intracellular signal transduction
positive regulation of GTPase activity
negative regulation of TORC1 signaling
GTPase activator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline