Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195555.1 | NP_001182484.1 | 10 | 1415 | ||
NM_001195556.1 | NP_001182485.1 | 10 | 1392 | |||
NM_014666.3 | NP_055481.1 | 10 | 1415 | |||
XM_011534701.2 | XP_011533003.1 | 10 | 1400 | |||
XM_017010087.1 | XP_016865576.1 | 10 | 1400 | |||
XM_017010088.1 | XP_016865577.1 | 10 | 1400 | |||
GenBank mRNA | AF434813.1 | - | 10 | 1210 | ||
AK092765.1 | - | 10 | 1362 | |||
AK300257.1 | - | 10 | 1383 | |||
BC004467.2 | - | 10 | 1276 | 3831533 | ||
BC013091.1 | - | 3 | 226 | |||
BC030264.1 | - | 2 | 1371 | |||
D79993.1 | - | 10 | 1311 | |||
KU178486.1 | - | 10 | 1210 | |||
KU178487.1 | - | 10 | 1210 |