Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014674.2 | NP_055489.1 | 1, 2 | 645 | ||
GenBank mRNA | AK292643.1 | - | 1, 2 | 513 | ||
AK298511.1 | - | 2, 3 | 177 | |||
BC019088.1 | - | 1, 2 | 652 | 4863779 | ||
D86967.1 | - | 1, 2 | 571 |