Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012291.4 | NP_036423.4 | 10 | 2216 | ||
XM_006719705.3 | XP_006719768.1 | 10 | 2283 | |||
XM_011539024.2 | XP_011537326.1 | 10 | 2244 | |||
XM_011539025.2 | XP_011537327.1 | 10 | 2255 | |||
XM_017020253.1 | XP_016875742.1 | 10 | 2334 | |||
GenBank mRNA | AY455930.1 | - | 9 | 2125 | ||
BC047603.1 | - | 8 | 1014 | |||
D79987.1 | - | 10 | 2263 |