Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142610.1 | NP_001136082.1 | 24 | 3165 | ||
NM_014683.3 | NP_055498.3 | 24 | 3165 | |||
XM_011524087.2 | XP_011522389.1 | 20 | 2462 | |||
XM_017025424.1 | XP_016880913.1 | 24 | 3053 | |||
XM_017025425.1 | XP_016880914.1 | 24 | 3053 | |||
XM_017025426.1 | XP_016880915.1 | 24 | 2972 | |||
XM_017025427.1 | XP_016880916.1 | 22 | 2507 | |||
XM_017025428.1 | XP_016880917.1 | 22 | 2426 | |||
GenBank mRNA | AB014523.1 | - | 24 | 3156 | ||
AK098316.1 | - | 23 | 2790 | |||
AK314345.1 | - | 24 | 2688 | |||
BC034988.1 | - | 25 | 2730 | 5268025 |