Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014704.3 | NP_055519.1 | 14 | 2245 | ||
XM_005244815.4 | XP_005244872.1 | 15 | 2345 | |||
XM_011542473.2 | XP_011540775.1 | 15 | 2545 | |||
XM_011542474.2 | XP_011540776.1 | 14 | 2263 | |||
XM_011542475.2 | XP_011540777.1 | 15 | 2371 | |||
XM_011542476.2 | XP_011540778.1 | 14 | 2347 | |||
XM_011542477.2 | XP_011540779.1 | 14 | 2173 | |||
XM_011542478.1 | XP_011540780.1 | 15 | 2545 | |||
XM_017002917.1 | XP_016858406.1 | 15 | 2353 | |||
XM_017002918.1 | XP_016858407.1 | 14 | 2071 | |||
XM_017002919.1 | XP_016858408.1 | 13 | 2025 | |||
GenBank mRNA | AB011134.1 | - | 14 | 2199 | ||
AK302190.1 | - | 5 | 933 | |||
BC001640.2 | - | 14 | 2210 |