Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014706.3 | NP_055521.1 | 17 | 2640 | ||
XM_005269241.4 | XP_005269298.1 | 17 | 2707 | |||
XM_011539026.2 | XP_011537328.1 | 17 | 2175 | |||
GenBank mRNA | AB020880.1 | - | 17 | 2424 | ||
AB073647.1 | - | 2 | 77 | |||
AF387506.1 | - | 17 | 2408 | |||
AK290209.1 | - | 17 | 2432 | |||
AK302332.1 | - | 11 | 1251 | |||
BC093784.1 | - | 17 | 2410 | 7939629 | ||
BC111983.1 | - | 17 | 2410 | 8327451 | ||
BC143253.1 | - | 16 | 2302 | 9051755 | ||
BQ231651.1 | - | 2 | 435 | |||
CR933631.1 | - | 10 | 7171 | |||
D63879.1 | - | 17 | 2405 |