Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014706.3 | NP_055521.1 | 18 | 2812 | ||
XM_005269241.4 | XP_005269298.1 | 18 | 2879 | |||
XM_011539026.2 | XP_011537328.1 | 18 | 2347 | |||
GenBank mRNA | AB020880.1 | - | 18 | 2596 | ||
AB073647.1 | - | 3 | 249 | |||
AF387506.1 | - | 18 | 2580 | |||
AK290209.1 | - | 18 | 2604 | |||
AK302332.1 | - | 12 | 1423 | |||
BC093784.1 | - | 18 | 2582 | 7939629 | ||
BC111983.1 | - | 18 | 2582 | 8327451 | ||
BC143253.1 | - | 17 | 2474 | 9051755 | ||
BQ231651.1 | - | 3 | 607 | |||
CR933631.1 | - | 11 | 7343 | |||
D63879.1 | - | 18 | 2577 |