Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
siRNA ID
HSS114607
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
7
954
NM_001204145.2
NP_001191074.1
5
646
NM_001204146.2
NP_001191075.1
5
824
NM_001204147.2
NP_001191076.1
4
612
NM_001204148.2
NP_001191077.1
4
621
NM_001321868.1
NP_001308797.1
6
894
NM_001321869.1
NP_001308798.1
6
903
NM_001321870.1
NP_001308799.1
6
894
NM_001321871.1
NP_001308800.1
6
909
NM_001321872.1
NP_001308801.1
6
903
NM_001321873.1
NP_001308802.1
6
903
NM_001321874.1
NP_001308803.1
5
824
NM_001321875.1
NP_001308804.1
5
830
NM_001321876.1
NP_001308805.1
5
824
NM_001321877.1
NP_001308806.1
5
824
NM_001321878.1
NP_001308807.1
5
815
NM_001321879.1
NP_001308808.1
5
824
NM_001321884.1
NP_001308813.1
5
824
NM_001321885.1
NP_001308814.1
5
839
NM_001321886.1
NP_001308815.1
6
874
NM_001321887.1
NP_001308816.1
6
714
NM_001321888.1
NP_001308817.1
5
646
NM_001321889.1
NP_001308818.1
6
883
NM_001321890.1
NP_001308819.1
5
815
NM_001321891.1
NP_001308820.1
5
815
NM_001321893.1
NP_001308822.1
6
712
NM_001321894.1
NP_001308823.1
6
721
NM_001321895.1
NP_001308824.1
5
655
NM_001321896.1
NP_001308825.1
6
993
NM_001321897.1
NP_001308826.1
6
721
NM_001321898.1
NP_001308827.1
5
670
NM_001321899.1
NP_001308828.1
6
721
NM_001321900.1
NP_001308829.1
5
815
NM_001321901.1
NP_001308830.1
5
655
NM_001321902.1
NP_001308831.1
5
655
NM_014707.3
NP_055522.1
5
646
NM_058176.2
NP_478056.1
4
501
NM_178423.2
NP_848510.1
5
646
NM_178425.3
NP_848512.1
4
510
XM_011515629.2
XP_011513931.1
5
604
XM_011515631.2
XP_011513933.1
6
918
XM_011515634.2
XP_011513936.1
5
604
XM_011515635.2
XP_011513937.1
6
1470
XM_011515636.2
XP_011513938.1
5
1191
XM_011515637.2
XP_011513939.1
5
615
XM_011515638.2
XP_011513940.1
5
1185
XM_011515639.2
XP_011513941.1
5
1382
XM_011515641.2
XP_011513943.1
6
1467
XM_011515642.2
XP_011513944.1
4
2196
XM_011515644.2
XP_011513946.1
6
1442
XM_011515645.2
XP_011513947.1
4
2181
XM_011515646.2
XP_011513948.1
4
2172
XM_011515648.2
XP_011513950.1
5
1200
XM_011515649.2
XP_011513951.1
5
1404
XM_011515651.1
XP_011513953.1
5
576
XM_017012826.1
XP_016868315.1
5
1176
XM_017012827.1
XP_016868316.1
6
1259
XM_017012828.1
XP_016868317.1
5
1384
XM_017012829.1
XP_016868318.1
5
585
XM_017012830.1
XP_016868319.1
5
576
XM_017012831.1
XP_016868320.1
5
585
XM_017012832.1
XP_016868321.1
5
585
GenBank mRNA
AB018287.1
-
5
610
AF124924.1
-
2
168
AJ459808.1
-
5
610
AK097178.1
-
5
604
AK296214.1
-
5
556
AK297404.1
-
5
600
AK304298.1
-
7
947
AK304410.1
-
4
575
AK316109.1
-
5
775
AY032737.1
-
4
460
AY032738.1
-
4
460
AY197371.1
-
4
469
BC111735.1
-
4
468
BC150328.1
-
5
621
8860495
BC152405.1
-
5
610
8862508
CR749550.1
-
4
577
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline